Learn More
Fisherbrand™ Flexaframe™ Vertical Mounting Supports
Great for glassware setups, etc., in small laboratories
Catalog No. 14-666-5Q
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
View alternate product
This item is not returnable. View return policy
Description
Description
Contents listed in “Contents of Flexaframe Sets” chart.
  • With wide, 18 in. (46cm) base extending out from lattice for maximum stability
  • Base can be permanently secured to benchtop with screws
  • Frame is 24L x 24 in.H (61 × 61cm)
Specifications
Specifications
18 in.
46 cm
Eight 2 in. rods, two 18 in. rods, eight 24 in. rods, 22 hook connectors, 4 supporting feet
Vertical Mounting Supports Set 1
Product Suggestions
Product Suggestions