Learn More
Fisherbrand™ Flexaframe™ Large-Capacity Supports
Excellent for complex glassware setups
Catalog No. 14-666-7Q
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
Description
Description
Excellent for complex glassware setups
  • 48 x 48 in. (122 x 122cm) frame
  • Base is 18 in. wide (46cm) for stability
Specifications
Specifications
18 in.
Ten 2 in. rods, three 18 in. rods, two 36 in. rods, ten 48 in. rods, 42 hook connectors, 6 supporting feet
Complex glassware setups, castaloy clamps and holders, disassemble in minutes
46 cm
Large Capacity Supports Set 3
Product Suggestions
Product Suggestions