Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fisherbrand™ Fingertip Timer

“Always at your fingertips”.
Includes: Magnetic back with Velcro™ strips for mounting, clock, stop watch, Two-year silver-oxide battery
Description
- Easy-to-use, three-button operation
- Counts down from 99 minutes, 59 seconds, in one-second increments
- Alarm may be silenced manually or turns off automatically after 30 seconds
- Memory retains last programmed time automatically for faster repeat testing
Order Info
Dimensions: 1.8 × 3.5 × 0.5 in. (4.6 × 8.9 × 1.3cm); weight: 1.2 oz. (34g)
Certifications
Individually serial-numbered Traceable™ Certificate is provided from an ISO 17025 calibration laboratory accredited by A2LA™. It indicates traceability to standards provided by NIST™.
Specifications
Fingertip Timer | |
0.0001 | |
LCD | |
0.5 x 1.81 x 3.5 in. (1.27 x 4.59 x 8.89 cm) | |
1.27 cm | |
4.59 cm | |
0.37 in. | |
0.93 cm | |
34.01 g | |
NIST |
99 min. 59 sec. | |
1 sec. | |
Silver Oxide | |
0.5 in. | |
1.81 in. | |
3.5 in. | |
8.89 cm | |
1.2 oz. | |
Magnetic back with Velcro™ strips for mounting, clock, stop watch, Two-year silver-oxide battery |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title