Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fisherbrand™ accumet™ AB150 pH Meters
For laboratories that need basic, accurate, easy-to-use pH/mV meter with exceptionally high accuracy and resolution.
Includes: AB150 Kit: Includes meter, TRIS compatible pH/ATC electrode, electrode arm, RS-232 and USB cables, 100/240V power supply, and manual
AB150 Meter only: Includes meter, electrode arm, RS-232 and USB cables, 100/240V power supply, and manual
Description
Intuitive, simple operation and high accuracy in a compact, affordable meter.
- Easy-to-understand prompts
- Graphic display with adjustable backlight
- Calibrate with custom pH buffer values
- Date and Time to meet GLP
- Password protection
- Optional calibration reminder and high/low alarm
- 500 point memory – log manually or at intervals
- Easily view calibration data
- 0.001 pH resolution and ±0.002 pH accuracy
pH Mode
- Range: -2.000 to +20.000 pH
- Resolution: 0.1/0.01/0.001 pH
- Accuracy: ±0.002 pH + 1 LSD
- Buffer Sets: USA: 2.000, 4.010, 6.997, 10.013, 12.000
- NIST: 1.678, 4.010, 6.865, 9.184, 12.460
- DIN: 1.090, 3.060, 4.650, 6.790, 9.230, 12.750
- FSCI: 1.000, 3.000, 6.000, 8.000, 10.000, 13.000
- CUSTOM: any two to five values, ≥1.0 pH unit apart
- Slope Display: up to five different slopes with offset
- Temp. Compensation: automatic or manual (0° to 100°C/32° to 212°F)
- RS-232 (phono plug), mini-B USB, stirrer
- English, French, Spanish, German, Italian, Chinese, Korean, and Portuguese
- 500 data sets, viewable
- Manual, timed (selectable every 3 to 3600 seconds)
- Printer or CSV format
- User selectable from 8 hours, 16 hours and 1 to 31 days
- User selectable, visual and audible, parameter based
Certifications
Select models available with Certificate of NIST Traceability
Warranty and Services
Meters: Three years parts and labor; Electrodes (included with meter kits): One year parts and labor
Specifications
±0.3°C | |
±0.002 | |
RS232 (phono plug), mini-B USB, stirrer | |
0.1/0.01/0.001 | |
±2000 mV | |
0.1°C | |
500 | |
NIST, ISO, FDA, USDA, EPA | |
AB150 pH Meter only | |
100/240 V |
±0.2 mV | |
Manual, timed (selectable every 3 to 3600 sec.), printer or CSV format | |
-2 to 20 | |
Up to 6 preset or 5 custom | |
0.1 mV | |
LCD | |
BNC, ATC | |
3 Years | |
pH Meter |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction