Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fisherbrand™ accumet™ AB150 pH Meters
For laboratories that need basic, accurate, easy-to-use pH/mV meter with exceptionally high accuracy and resolution.
Includes: AB150 Kit: Includes meter, TRIS compatible pH/ATC electrode, electrode arm, RS-232 and USB cables, 100/240V power supply, and manual
AB150 Meter only: Includes meter, electrode arm, RS-232 and USB cables, 100/240V power supply, and manual
Description
Intuitive, simple operation and high accuracy in a compact, affordable meter.- 0.001pH resolution
- Calibrate with user-selected custom pH buffer values
- Optional stirring probe accessory (Cat. No. 13-620-BSP) with five speed settings
- Compact size conserves valuable bench space
- Large display shows measurement and temperature at all times
- Automatic Temperature Compensation (ATC) in °C or °F
- Graphic display with adjustable backlight
- Date and Time to meet GLPPassword protection
- Optional calibration reminder and high/low alarm
- 500-point memory — log manually or at intervals
- Easy-to-understand prompts
- Easily view calibration data
- 100 to 240V power supply includes multi-plugs for use around the globe
- Select preferred language as displayed on instrument — choose from English, German, Chinese, French, Italian and Spanish
Certifications
Select models available with Certificate of NIST Traceability
Warranty and Services
Meters: Three years parts and labor; Electrodes (included with meter kits): One year parts and labor
Specifications
±0.3°C | |
±0.002 | |
RS232 (phono plug), mini-B USB, stirrer | |
0.1/0.01/0.001 | |
±2000 mV | |
0.1°C | |
500 | |
NIST, ISO, FDA, USDA, EPA | |
AB150 pH Meter Kit | |
100/240 V |
±0.2 mV | |
Manual, timed (selectable every 3 to 3600 sec.), printer or CSV format | |
-2 to 20 | |
Up to 6 preset or 5 custom | |
0.1 mV | |
LCD | |
BNC, ATC | |
3 Years | |
pH Meter Kit |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction