Learn More
Fisherbrand™ 6-Place Fixed Angle Rotor for Horizon mini-E
Description
For tubes up to 17 × 125mm
Specifications
6-place Fixed Angle Rotor | |
3,900 rpm | |
1.7 cm | |
Fixed |
1,857 x g | |
12 cm | |
Horizon Mini E Centrifuge (22-029375) |