Learn More
Fisher Chemical™ HistoPrep™ Frozen Tissue Embedding Media
Description
- A special formulation of water-soluble glycols and resins used as a medium for frozen sectioning
- Provides a solid bond with object holder to reduce tissue dislodging by knife pressure
Specifications
Clear | |
Paraffin | |
125mL | |
Used as a medium for frozen sectioning |