Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fisher BioReagents™ Thrombin, From Bovine Plasma
Specifications
Color | White |
---|---|
For Use With (Application) | Clotting factor tests, coagulation assays and the defibrination of Blood or Plasma |
Content And Storage | <0°C |
Source | Bovine Plasma |
Product Type | Thrombin |
Catalog Number | Quantity | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Quantity | Price | Quantity & Availability | ||||||
BP25432
|
2 kU |
Item Discontinued
This item has been discontinued by the supplier. Please Sign In to view product availability in your area. View alternate product |
N/A | ||||||
BP254310
|
10 kU |
Item Discontinued
This item has been discontinued by the supplier. Please Sign In to view product availability in your area. View alternate product |
N/A | ||||||
BP254325
|
25 kU |
Item Discontinued
This item has been discontinued by the supplier. Please Sign In to view product availability in your area. View alternate product |
N/A | ||||||
Description
- Application: Thrombin is used for clotting factor tests, coagulation assays, and the defibrination of blood or plasma
- Storage: Store below 0°C
Specifications
White | |
<0°C | |
Thrombin |
Clotting factor tests, coagulation assays and the defibrination of Blood or Plasma | |
Bovine Plasma | |
Solid |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title