Learn More
Fisher BioReagents™ SurePrep™ Small RNA Purification Kit
Optimized for enrichment of small RNA by removing large RNA for studying regulation of gene expression
Includes: Lysis solution (2 x 20mL), Wash solution (22mL), RNA elution buffer (6mL), Large RNA removal columns (25), Small RNA enrichment columns (25), Collection tubes (50), Elution tubes (1.7mL) (50) and Product insert
Description
- Isolate all small RNA molecules (<200 nucleotides)
- Minimal contamination from large RNA molecules and genomic DNA
- Fast and easy processing using rapid spin-column format
- No phenol or chloroform extractions
- High quality small RNA such as miRNA and siRNA can be used in various downstream applications
Applications:
Purified small RNA is suitable for use in RT-PCR, Northern blotting and microarray analysis
Specifications
Spin Column | |
Lysis solution 2 x 20mL, wash solution 22mL, RNA Elution Buffer 6mL, large RNA removal columns 25, small RNA enrichment columns 25, collection tubes 50, elution tubes 1.7mL 50 and product insert | |
15μg HeLa cells (1 x 106 cells), 50μg E. coli (1 x 109 cells) |
RT-PCR, northern blotting and microarray analysis | |
25 Preps |