Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fisher BioReagents™ NZY Broth (Powder/Molecular Genetics)
Specifications
Content And Storage | Keep refrigerated. (Store below 4°C/39°F.) Store in a cool, dry, well-ventilated area. |
---|---|
Form | Powder |
Grade | Molecular genetics |
Media Type | NZY Broth |
Product Type | Microbiology Media |
Catalog Number | Quantity | Packaging Type | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Quantity | Packaging Type | Price | Quantity & Availability | |||||
BP2465100
|
100 g | Poly Bottle | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | |||||
BP2465500
|
500 g | Poly Bottle | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | |||||
BP24652
|
2 kg | Poly Bottle | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | |||||
Description
Specifications
Keep refrigerated. (Store below 4°C/39°F.) Store in a cool, dry, well-ventilated area. | |
Molecular genetics | |
Microbiology Media |
Powder | |
NZY Broth |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title