Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fisher BioReagents™ LB Agar, Lennox (Granulated)
Used as a solidifying agent to provide the solid medium to support the growth of microbes
Specifications
Form | Granulated Powder |
---|---|
Product Type | Agar |
For Use With (Application) | Used as a solidifying agent to provide the solid medium to support the growth of microbes |
Catalog Number | Packaging Type | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Packaging Type | Quantity | Price | Quantity & Availability | |||||
BP97452
|
Poly Pail | 2 kg | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | |||||
BP9745500
|
Poly Bottle | 500 g | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | |||||
Description
- Granulated LB Agar, Lennox media
- CAS: [91079-40-2], [8013-01-2], [7647-14-5], [9002-18-0]
- Store at room temperature
Specifications
Granulated Powder | |
Agar | |
Used as a solidifying agent to provide the solid medium to support the growth of microbes |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title