Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fisher BioReagents™ Bovine Serum Albumin, Heat Shock Treated
Suitable for immunological studies
$427.00 - $2640.00
Specifications
For Use With (Application) | Immunological Applications |
---|---|
pH Range | 7 ±0.3 |
Source | Bovine |
Packaging | Poly Bottle |
Catalog Number | Quantity | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Quantity | Price | Quantity & Availability | ||||||
BP1600-100
|
100g |
Each for $427.00
Case of 6 EA for $2,050.00
Order in Bulk
Request bulk or custom formats |
|
||||||
BP1600-1
|
1kg |
Each for $2,640.00
Case of 4 EA for $8,400.00
Order in Bulk
Request bulk or custom formats |
|
||||||
Description
CAS: 9048-46-8
Specifications
Immunological Applications | |
7 ±0.3 | |
Bovine | |
Poly Bottle |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title