Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Labconco™ FilterMate™ Portable Exhausters
The FilterMate uses HEPA filter, carbon filter or combination of both filters to remove hazardous powders, particulates or vapors from exhaust air stream and return filtered air to environment.
Supplier: Labconco™ 3970003/EMD
Description
- Compact, corrosion-resistant housing of glacier white and light gray powder-coated steel with rubber feet
- Variable-speed, non-sparking motorized impeller
- Oversized HEPA filter, efficient impeller and perforated exhaust cover work together to provide lower system exhaust velocity and noise pressure levels
- Inherently safe design via negative pressure plenum that surrounds positive pressure impeller so that if leak occurs, unfiltered air is captured and refiltered
- Front-mounted rocker switch to control power to blower
- Rear-mounted, auxiliary electrical outlet that may be connected to airflow monitor or other user-supplied accessories
- Only one FilterMate is required for all 2 ft., 3 ft. and 4 ft. wide enclosures, Multiple FilterMates may be used for 5 ft., 6 ft. and 8 ft. wide enclosures
- IEC three-wire electrical cord 1.9m (6.5 ft.) with plug included (115V models)
- Models with HEPA filters have a true bag-in/bag-out filter disposal system for safely removing and replacing filter without detaching hose
- HEPA filter may be easily accessed by removing exhaust cover for field leak testing by certified technician
- Thimble connection (included with some models) provides added safety by connecting FilterMate to existing house exhaust system
Certifications
UL 61010-1; CAN/CSA C22.2 No. 61010.1
Warranty and Services
1 year
Specifications
Exhauster | |
Exhaust connection: None, room exhaust | |
22.7 in. | |
60 Hz |
HEPA | |
200 cfm (60 to 100 fpm) | |
57.6 cm | |
115 V |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction