Learn More
Labconco™ Filtered Fume Hood Accessory, HEPA Filter
SDPSCI_ED
For use with Protector Echo and Airo Filtered Fume Hood
Supplier:  Labconco™ 9577401/EMD
Catalog No. S28324
Add to Cart
Description
Description
  • Optional HEPA filter can be placed between prefilter and main level Neutrodine filter to provide capture of non-hazardous particulates
  • Efficiency: 99.995%, H14 type
  • Number of filters needed depends on hood's width
Specifications
Specifications
HEPA Filter
76.2 cm
4 in.
16 in.
30 in.
Protector™ Echo™ and Airo™ Filtered Fume Hood
10.1 cm
40.6 cm