Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fisherbrand™ FilterCart Advanced+ Kit
FilterCart Advanced+ is a light-weight, portable and easy to maneuver dust and fume extractor with a warning signal when the filter is full.
This item is not returnable.
View return policy
Description
- FilterCart Advanced+ is a light-weight, portable and easy to maneuver dust and fume extractor with a warning signal when the filter is full.
- It includes an easy-to-position.
- Original extraction arm with 10 ft. long arm.
- The arm is already fitted to the unit enabling a fast and easy setup.
Specifications
Filter Cart | |
PVC | |
1145 cm | |
1140 cm | |
770 cm |
Blue | |
45 in. | |
55.5 in. | |
30 in. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction