Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Agilent BioTek Filter Cube, CFP FRET V2
For use with microplate readers and imagers
Supplier: Agilent BioTek 1225118
Description
- Accessory for Agilent BioTek instruments range of microplate washers, readers and cell imagers
- Fluorescence filter
- Requires 1225011 LED cube
Specifications
Microplate readers and imagers |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction