Learn More
Quidel Corporation MicroVue Human FGF-23 (Intact) ELISA Kit
Supplier: Quidel Corporation 606600
The MicroVue Human FGF-23 (Intact) ELISA Kit is for the measurement of FGF-23 in plasma or cell culture media.