Learn More
Thermo Scientific Beta Cyanoethyl Fastphoramidite, dG(dmf), 330628-04-1, MFCD09842106, 0.5 g
Supplier: Thermo Scientific 401182
MF: C43 H53 N8 O7 P, MW: 824.915, Chemical Name: dG(dmf) beta CEP. The dG(dmf) Beta Cyanoethyl Fastphoramidite is used as a component of a fast deprotecting chemistry set including dA(Bz), dC(Bz) and T. FastPhoramidite series deprotects in 1 hour at 65°C.