Learn More
Thermo Scientific Beta Cyanoethyl Phosphoramidite, dG(iBu), 93183-15-4, MFCD00055147, 2 g
Supplier: Thermo Scientific 401161
MF: C44 H54 N7 O8 P, MW: 839.926, Chemical Name: dG(iBu) beta CEP. The dG(iBu) Beta Cyanoethyl Phosphoramidite is used as a component of a deprotecting chemistry set including dA(Bz), dC(Bz) and T. Standard series of phosphoramidite deprotects in 6-8 hours at 55°C.