Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Gibco™ Fetal Bovine Serum, qualified, USDA-approved regions
Important Update: We've recently introduced Value Fetal Bovine Serum (FBS) specification-based products. These improved products are alternatives to our Qualified USDA origin and Brazil (South American) origin SKUs. Product stability, performance, manufacturing, storage, and transport processes are unchanged.
Supplier: Gibco™ A3160602
Description
Sera category: Standard- Sera for cell culture with robust cell lines
- Good value for basic research
- Endotoxin level: ≤ 50 EU/mL (levels routinely ≤ 10 EU/mL)
- Hemoglobin level: ≤ 25 mg/dL
- Origin: Countries meeting USDA importation requirements
- ISO 13485-certified, processed in FDA registered facilities
- Labeled for in vitro diagnostics (compliant with highest level of USP sterility testing)
- Triple filtered at 0.1 μm
- Gibco™ bottle is easier to use in the hood, reduces the risk of contamination and helps you perform cell culture more consistently
- Peel-off sticker on label provides handy reference for lot number and expiration date recording in lab notebook
Aliquot-free 50 mL bottle
The Gibco™ One Shot™ 50 mL bottle is designed to eliminate the need to aliquot, and the risk of contamination and variability that comes with it, improving your chances for consistent results.
Features include:
- Wide mouth, chamfered edge, and curved shape for easy pouring and pipetting
- Engineered to freeze/thaw without splitting or breaking
- Fits in test tube racks and frees up freezer space
- No labeling mishaps with our easy-to-read package label containing all of the information you need
- 33% less waste generated than aliquoting from 500 mL bottles
For Research Use or Further Manufacturing Use only. Serum and blood proteins are not for direct administration into humans or animals.
Order Info
Shipping Condition: Dry Ice
Specifications
Storage conditions: ≤-10°C Shipping conditions: Frozen |
|
Bovine | |
Liquid | |
Fetal Bovine Serum | |
Standard (Sterile-filtered) | |
Triple-filtered, 0.1 μm | |
Qualified | |
Frozen |
Fetal Bovine Serum, qualified, One Shot™ format, USDA-approved regions | |
Mexico | |
One Shot | |
10 x 50 mL | |
Sterile | |
Fewer resources used and less waste | |
One Shot™ |
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction