Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Gibco™ Fetal Bovine Serum, qualified, Australia
Important Update: We've recently introduced Premium Plus Fetal Bovine Serum (FBS) specification-based products. These improved products are alternatives to our Certified US origin, New Zealand origin, and Qualified Australia orgin SKUs. Product stability, performance, manufacturing, storage, and transport processes are unchanged.
Supplier: Gibco™ A3161001
Description
If you’re one of the more than 80% of researchers who aliquots their FBS into 50mL conical tubes, your life is about to change for the better. Transferring your FBS takes away your control — of how you spend your time, your contamination risk, and your volume consistency. It also wastes costly material.The new Gibco™ One Shot™ FBS 50mL bottle design eliminates the need to aliquot, and the variability that comes with it. And that improves your chances for consistent results.
Product Features:
- Wide mouth, chamfered edge, and curved shape for easy pouring and pipetting
- Engineered to freeze/thaw without splitting or breaking
- Fits in test tube racks and frees up freezer space
- No labeling mishaps with our easy-to-read package label has all the information you need
- Generates 33% less waste than aliquoting from 500mL bottles
Order Info
Shipping Condition: Dry Ice
Specifications
Storage conditions: ≤-10°C Shipping conditions: Frozen |
|
Fetal | |
Australia | |
One Shot | |
50 mL | |
Sterile | |
Fewer resources used and less waste | |
Qualified, Cell culture grade | |
Frozen |
Fetal Bovine Serum, qualified, One Shot™ format, Australia | |
Bovine | |
Liquid | |
Fetal Bovine Serum | |
Standard (Sterile-filtered) | |
≤5 EU/mL | |
≤30 mg/dL | |
One Shot™ |
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction