Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Corning™ Fetal Bovine Serum, Premium, New Zealand Origin
Collected at government registered facilities in New Zealand
Supplier: Corning™ 35108CV/RSD
This item is not returnable.
View return policy
Description
- Exported from the country of origin accompanied with a Veterinarian Health Certificate and imported under a USDA-APHIS issued import permit.
- True pooled and is triple 0.1 micron sterile filtered that passes sterility, mycoplasma, and 9CFR viral testing requirements.
- Validated process used to heat inactivate serum at 56°C ± 2°C for 30 minutes via bulk inactivation of entire lot in the pooling tank or individual bottles in a water bath.
Specifications
-40°C to -10°C | |
New Zealand | |
500 mL | |
Sterile | |
Yes |
Bovine | |
Liquid | |
Heat-inactivated, Gamma-irradiated | |
New Zealand Origin |
Safety and Handling
ShelfLife : 60 months
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction