Learn More
Gibco™ Fetal Bovine Serum (FBS), Premium, heat-inactivated
![GREENER_CHOICE](/content/dam/fishersci/glyphs/greener_choice.png)
Gibco fetal bovine sera offer excellent value for basic cell culture, specialty research, and specific assays, earning the trust of researchers with consistent quality and award-winning support that helps meet your research needs and budget requirements.
$95.00 - $898.00
Specifications
Content And Storage | Store at ≤-10°C. |
---|---|
Age | Fetal |
Species | Cattle/Bovine |
Country of Origin | Refer to CoA for specific origin |
Form | Frozen |
Catalog Number | Mfr. No. | Quantity | Format | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Format | Price | Quantity & Availability | ||||
A5670501
![]() ![]() |
Gibco™
A5670501 |
50 mL | One Shot |
Each for $95.00
|
|
||||
A5670502
![]() ![]() |
Gibco™
A5670502 |
10 x 50 mL | One Shot |
Pack of 10 for $898.00
|
|
||||
A5670801
![]() |
Gibco™
A5670801 |
500 mL | Bottle |
Each for $760.00
|
|
||||
Description
Gibco fetal bovine sera offer excellent value for basic cell culture, specialty research, and specific assays, earning the trust of researchers with consistent quality and award-winning support that helps meet your research needs and budget requirements.
Gibco sera: trusted quality for consistent performance
Sera category: Premium
- Use for general cell culture with common cell lines
- Low endotoxin and hemoglobin levels
- Endotoxin level: <10 EU/mL
- Hemoglobin level: <25 mg/dL
- Origin: refer to CoA for specific origin. All products may not be available in all regions due to importation regulations.
- US Department of Agriculture (USDA) safety tested for the absence of bluetongue and akabane from countries that are USDA-approved for importation into the US
Gibco serum delivers
- ISO 13485–certified; processed in FDA registered facilities
- Triple-filtered at 0.1 μm
- Gibco bottle easier to use in hood; helps reduce risk of contamination and improve consistency
- Peel-off sticker on label provides handy reference for lot number and expiration date recording in lab notebook
Specifications
Store at ≤-10°C. | |
Cattle/Bovine | |
Frozen | |
Heat-inactivated | |
Triple-filtered, 0.1 μm | |
Up to 60 quality tests including 9CFR virus testing, endotoxin, and performance | |
Less waste | |
Premium |
Fetal | |
Refer to CoA for specific origin | |
Fetal Bovine Serum | |
Sterile-filtered | |
Premium | |
≤10 EU/mL | |
≤25 mg/dL | |
Dry Ice |
Safety and Handling
ShelfLife : 5 Years
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.