Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3B Scientific™ Feline Heart and Lung Model
SCI_ED

3B Scientific Feline Heart And Lung Model demonstrates an infestation of a single heartworm (dirofilaria immitis).

Supplier:  3B Scientific™ 1019584

Catalog No. S00595


Add to Cart
This item is not returnable. View return policy

Description

Description

  • Average feline heart and lungs infested with a single heartworm (dirofilaria immitis).
  • Cut-away view demonstrates structures in the heart (right ventricle, pulmonary trunk, pulmonary arteries) and lungs where adult-stage heartworms are normally found. The cut section of the lung indicates patches of heartworm-induced irritation.
    • Model size: 3.75 x 2.75 x 2.5 in.
    • Base: 6.5 x 5 in.
Specifications

Specifications

Feline Model
6.5 x 5 in. Base
Heart, Lungs, Single Heartworm
3.75 x 2.75 x 2.5 in.
Anatomy Demonstrations
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit