Learn More
Medex Supply FEATHER SCAPELS 22 DISPOSABLE
Supplier: Medex Supply GRF2975#22
Size 22, Scalpel, Sterile, 20/bx Features heat-treated stainless steel blade securely integrated with rigid, chemically resistant plastic handle. Individually wrapped in hermetically-sealed aluminum foil. Blades are protected with snap-off cover. Scalpels available with choice of blad