Learn More
Shelby™ FDP™ Tanned Cowhide Leather Gloves - Knit Wrist
Catalog Number p-9490004
Encompass
Click to view available options
Size:
Small
This item is not returnable. View return policy
This item is not returnable. View return policy
Fire-retardant and heat-resistant heavyweight gloves meet the demands of firefighting
  • Durable outer shells are made of either soft, koala-tanned cowhide leather or abrasion-resistant brushed pigskin
  • Gloves feature Gore RT7100 glove barrier fabric laminated to SEF modacrylic fleece
  • Lining is sewn to shell at fingertips and wrist to eliminate pullouts
  • Seams are sealed with DuPont™ silicone sealant
  • Gunn cut with wing thumb construction
  • Welted thumb and fourchette seams
  • Glove body extends 2 in. (5 cm) above wrist crease with shirred elastic at wrist
  • 0.25 in. (6.3 mm) hanger loop for easy storage
Certifications/Compliance NFPA 1971, CE PPE, EN 659:1996, EN 388:1994
Color Blue
Material Cowhide Leather, Nomex
Product Type Structural Firefighting Gloves
Size Small
Glove Cuff Style Knit
Liner Material Thermal