Learn More
Lonza Walkersville FBM Fibroblast Growth Basal Medium, 500 mL
Supplier: Lonza Walkersville CC3131
FBM basal medium is a component sold separately. This is part of the FGM -2 and FGM -3 Fibroblast Growth Media BulletKits (Catalog Numbers CC-3132 and CC-4526, respectively) which consist of basal medium and supplements packaged together.FGM-2 Fibroblast Growth Medium-2 (CC-3132) was developed to support the growth of most primary human fibroblasts including lung and dermal fibroblasts. FGM-2 Medium contains 2% serum.FGM-3 Fibroblast Growth Medium-3 (CC-4526) was developed specifically for cardiac fibroblasts. FGM-3 Medium contains contains 10% serum. Purchase our Lung Fibroblast Cell Culture Kit (catalog no. CC-2512B) and get a discounted price on cells and media. The kit consists of a cryopreserved ampule of lung fibroblast cells (catalog no. CC-2512) and FGM-2 BulletKit Growth Media (catalog no. CC-3132).Content:1 x FBM Basal Medium, 500 mL
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.