Learn More
Cen-Med False Bottom Tubes
For use as a secondary tube for barcoding on automated analyzers.
Manufacturer: Cen-Med 1261543
Description
- Amber
- 16 x 95
Specifications
Amber | |
Beckman™, Corning™, Hitachi™, IA™, Kodak™, Olympus™, Ortho™, Tosoh™, Vitros™; and other analyzers. | |
1000/Pk. |
False Bottom Tube | |
Secondary tube for barcoding on automated analyzers. |