Learn More
Glas Col LLC Heating Mantle
SDP
Supplier:  Glas Col LLC 100A O856
Encompass_Preferred
Fabric mantle for 2L Squibb funnel, to 400 degrees C maximum internal operating temperature, 280W, 115V CSA
Catalog No. NC1837251
Due to product restrictions, please to purchase or view availability for this product.