Learn More
Fisherbrand™ Fabric Chair Chrome - Medium Bench Height with Medium Back, Seat Tilt, Adjustable Arms, Chrome Foot Ring, and Casters in Fabric
SDPSCI_ED
Fisherbrand™ Fabric Task Chairs in Chrome with Medium Back are height adjustable for 36 in. work surface and feature ergonomic adjustments, larger back, arms, chrome adjustable foot ring, and self-braking casters. Heavy-duty olefin fabric in a variety of colors will suit any work space.
 View more versions of this product
Catalog No. S36505
Add to Cart
Description
Description
  • Medium Back
  • Adjustable from 21.5-29 inches from the top of the seat to the floor
  • Features ergonomic seat height adjustment
  • Locking back tilt
  • 8-degree forward and 3-degree backward locking seat tilt
  • 6-way adjustable arm rests adjust up and down, in and out, and twist toward and away from user
  • 20-Inch Chrome adjustable foot ring with internal lock
  • Cast-aluminum base
  • Chrome-hooded, dual-wheel, self-braking, non-marring casters suitable for hard floors
  • Heavy-duty fabric
Specifications
Specifications
General Seating Chair
Polyurethane foam, Olefin
136 kg
Medium Bench Height
Round tube chrome foot ring with internal lock
20 in.
Yes
18 in.
Polished Cast Aluminum
Yes
Yes
2.5 in.
19 in.
46 lb.
Ergonomic 8 degree forward and 3 degree backward seat tilt locks in place
300 lb.
Standard
Chrome-hooded dual-wheel, self-braking, non-marring casters (suitable for both hard and carpeted floors)
Gray
Yes (Pneumatic)
21.5 to 29 in.
Yes
25 in.
Yes
15 year parts, lifetime on pneumatic lift, 5 year on upholstery
6-way adjustable (move up and down, in and out, and twist toward or away from the user)
Recommendations
Recommendations
Safety and Handling
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov