Learn More
Epredia™ Fa-Tach™ Series 815 Laser Slotted Cassettes with Attached Lids
Manufactured specifically for laser printing, the Fa-Tach™ Series 815 Laser Slotted Cassettes come with attached lids and are available in multiple colors.
Supplier: Epredia™ 8150918LM
This item is not returnable.
View return policy
Description
- Fa-Tech™ Laser Cassettes are designed for use on both the NOVA™ and VEGA™ Laser Printers
- Contains a special constituent chemical compound that carbonizes when exposed to the laser beam
- Each style comes in multiple different colors
- Each cassette style is designed to maximize fluid exchange during processing and come in stacks held together with tape for ease of loading into Fa-Tech™ cassette hoppers for printing
- Series 815 come in taped stacks of 40 cassettes with lids attached; 45 stacks per case for a total of 1800 cassettes
- The Fa-Tech™ Laser Cassette is not compatible with thermal tape style printers
Specifications
Slots | |
Yes | |
Lilac |
40 Cassettes/Taped Stack, 45 Stacks/Cs. | |
Attached Lid |