Learn More
LGC Biosearch Technologies EZ-Tn5 Transposase, 10 units @ 1 U/LA hyperactive form of the Tn5 transposase optimised for in vitro transposon mutagenesis (transposomics).
Supplier: LGC Biosearch Technologies TNP92110
EZ-Tn5™ Transposase is a hyperactive Tn5 transposase for random insertion of an EZ-Tn5 Transposon into any cloned DNA in vitro or for preparing an EZ-Tn5 Transposome for random insertion of a EZ-Tn5 Transposon into the genomic DNA of living bacteria.
This item is not returnable.
View return policy