Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PIP™ Extreme™ Work VizX™ Hi-Vis Orange Industrial Gloves with Suede Palms
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
Description
The engineered microfiber Tough-X™ suede palm of these gloves is fast drying and enhances grip in dry and light oil applications while providing dexterity. These gloves last up to 5 times longer than traditional synthetic leathers.
- S-patch on palm for durability and grip
- Reinforced fingertips and thumb crotch for extended glove life, enhanced comfort, increased abrasion resistance and added durability in a high-wear area
- Spandex fabric back for comfort and breathability
- Slip on cuff with hook and loop closure
- Touchscreen compatibility on index finger
- Hi-Vis coloring makes gloves very easy to see
- Launderable for extended life and reduced replacement costs
Specifications
Chlorinated | No |
Construction | Cut & Sewn |
Applications | Industrial |
Color | Hi-vis Orange |
Material | Spandex, Suede |
Product Type | Multi-activity Gloves |
Size | Small |
Gender | Men's |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction