Learn More
GENIE GROUP Mini Laser Photo Tachometer Counter
Supplier: GENIE GROUP 461920NIST
Extech's 461920 is a tachometer for RPM measurements. Make safe, non-contact RPM measurements (2 to 99,999 RPM) of rotating objects by pointing the integral laser (1.6-ft. target distance) at the reflective tape placed on the object to be measured. Complete with 9-Volt battery, reflective tape and user manual.