Learn More
Supply Solutions EX-CELL CHO Cloning Medium, 500mL
Supplier: Supply Solutions SIGMA C6366500ML
This animal component-free medium is designed to support clonal survival and growth of Chinese Hamster Ovary (CHO) cell lines, with results comparable to the traditional method using 10% fetal bovine serum.