Learn More
Thermo Scientific™ eVol™ XR Dispensing System Plungers
Order these plungers for use with the respective size syringe on the Thermo Scientific™ eVol™ Dispensing System.
Specifications
Product Type | Dispensing System Plunger |
---|---|
Product Line | HyperSep |
For Use With (Equipment) | Electronic Dispensing System |
Catalog Number | Mfr. No. | Description | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Description | Price | Quantity & Availability | |||||
66-002-041
|
Thermo Scientific™
66002041 |
Replacment plunger for 1000µL eVol system, 1 pk | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | |||||
66-002-030
|
Thermo Scientific™
66002030 |
Replacment plunger for 50µL eVol system, 1 pk | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | |||||
Description
Use these Thermo Scientific™ eVol™ Dispensing System Plungers with the respective size syringe on the eVol Dispensing System. Like the syringes, these plungers are available in 5 to 1000μL sizes. The unique, hand-held eVol Dispensing System accurately and reproducibly performs a wide variety of liquid handling procedures. The easy-to-use device couples an analytical syringe to a programmable electronic controller, enabling highly reproducible dispensing for all users.
The eVol Dispensing System is ideal for laboratories that need to repeatedly dispense nonaqueous liquids, offering greater accuracy and reproducibility for applications such as:- Routine dispensing of volatile solvents, hazardous, corrosive or viscous chemicals
- Preparation of calibration standards
- Addition of internal standards
Repeatable Performance for Higher Confidence in Your Results
- Can be calibrated easily by a user to ensure validity of results
- Intuitive, easy-to-use interface with touch wheel and full-color screen
- Complies with GLP and GMP guidelines
- Integrated XCHANGE coupling allows syringes to be quickly and easily changed, preventing possible cross-contamination
- Addition of Thermo Scientific™ MEPS™ (Micro Extraction by Packed Sorbent) technology to an XCHANGE syringe enables automated sample preparation by SPE in a hand-held system
Specifications
Dispensing System Plunger | |
HyperSep | |
Electronic Dispensing System |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.