Learn More
Chemglass Life Sciences 20L Flask, Evaporating, Large Scale, 100mm Flange, 435mm OAH
Supplier: Chemglass Life Sciences CG133706
20L Flask, Evaporating, Large Scale, 100mm Flange, 435mm OAH. Flasks are compatible with Buchi large scale rotary evaporators. Flasks to fit large scale rotary evaporators. Flasks have a HEAVY WALL for vacuum applications and are balanced to prevent run-out during rotation.