Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ethyl Acetate (Certified ACS), Fisher Chemical™
Click to view available options
Quantity:
500 mL
1 L
4 L
20 L
200 L
Packaging:
Aluminum Can
Glass Bottle
Safe-Cote™ Amber Glass
Steel Drum
Steel Pail
Specifications
Melting Point | -83.5°C |
Color | Colorless |
Density | 0.902g/cm³ |
Boiling Point | 75°C |
Quantity | 4 L |
Identification | Pass Test |
Packaging | Safe-Cote™ Amber Glass |
Physical Form | Liquid |
Assay Percent Range | ≥99.5 % |
Residue after Evaporation | 0.003% max. |
Show More |
Safety and Handling
Hazard Category | DANGER! |
Hazard Statement | DANGER! |
Precautionary Statement | Emergency Overview Highly flammable. Vapors may cause drowsiness and dizziness. Repeated exposure may cause skin dryness or cracking. May cause central nervous system effects. May cause skin and respiratory tract irritation. May be harmful by inhalation, in contact with skin and if swallowed. Use only under a chemical fume hood. Use explosion-proof equipment. Wear personal protective equipment. Do not breathe vapors or spray mist. Do not get in eyes, on skin, or on clothing. Keep away from open flames, hot surfaces and sources of ignition. Keep away from open flames, hot surfaces and sources of ignition. Use only non-sparking tools. NFPA Health:1 Flammability:3 Instability:0 |
DOTInformation | DOT Class 3, : Flammable Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction