Learn More
Expression Systems ESF AF INS CELL CULT MED 20L
Supplier: Expression Systems 9930020
20L Media Transfer Bag
Expression Systems ESF AF is an animal origin-free formulation for insect cell culture. It has been thoroughly tested on Sf9, Sf21, Tni (High Five™) and S2 cell lines and is comparable to ESF 921 for growth and expression. ESF AF is the logical choice for further manufacturing under cGMP/FDA regulatory compliance and has a Drug Master File with the US FDA.
• 100% animal component-free
• Sf9 cell densities of 20 x 106 cells/ml or greater
• Ideal for scale-up manufacturing
• Works with a wide variety of baculovirus and host cell platforms
• Optimized for applications such as production of sub-unit vaccines, virus-like particles and gene therapy vectors (rAAV all serotypes)
• Available in 1L bottles and media transfer bags of 10, 20 and 50L volumes
• Complete, ready-to-use, contains L-glutamine
Always in stock for immediate shipment
Download Instructions for Use and/or COA please visit: www.expressionsystems.com
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.