Learn More
Expression Systems ESF 921 Insect Cell Culture Medium, Protein Free, 1L Bottle
Supplier: Expression Systems 9600101
ESF 921™ Insect Cell Culture Medium is a complete serum-free, protein-free medium developed for robust cell growth, protein production and baculovirus vector production for a wide range of insect cells including Sf9, Sf21, Tni (High Five™), and Drosophila S2. ESF 921 contains L-Glutamine and Kolliphor P188 (Pluronic® F68).This serum-free, protein-free insect cell culture medium provides unmatched cell growth and expression for a wide range of insect cells including Sf9, Sf21, Tni (High Five™) and Drosophila S2.• Sf9 cell densities of 15 x 106 cells/ml or greater• Produces high titer baculovirus vector yields of 5E8 to 3E9 infectious units/ml• Complete, ready-to-use, contains L-glutamine• Ideal for scale-up manufacturing• Works with a wide variety of baculovirus and host cell platforms• In stock for immediate shipment• Adapted cells availableDownload COA: www.expressionsystems.com. Kolliphor® and Pluronic® are registered trademarks of BASF SE.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.