Learn More
HTL-Strefa DropSafe ErgoLance Micro Flow Lancet
Sterile, single-use devices designed for the sampling of capillary blood from the fingertip of patients.
$6.08 - $1999.00
Specifications
Product Type Safety lancet
For Use With (Application) Lancet for blood testing
Material Medical sharp
Quantity 100 ct. per box
View More Specs
Products 3
Catalog Number Mfr. No. Color Needle Gauge Price Quantity & Availability  
Catalog Number Mfr. No. Color Needle Gauge Price Quantity & Availability  
22-192-021
View Documents
HTL-Strefa
8481
Green 30G
Pack of 100 for $6.08
Save 93%
Reg : $89.30
The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Learn More
Case of 20 PK for $121.60
Save 93%
Reg : $1,738.00
The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Learn More
Add to Cart
22-192-022
View Documents
HTL-Strefa
8482
Blue 25G
Case of 2000 for $1,869.00
Add to Cart
22-192-023
View Documents
HTL-Strefa
8483
Pink 21G
Case of 2000 for $1,999.00
Add to Cart
Description
Description
  • Special lancet shape distracts attention from needle stick
  • Concave housing for comfortable finger recess maximizes economy
  • No release button
  • Gentle pressure activation
  • Ribbed protective cap guarantees needle sterility and gives a secure grip for easy twist-off removal
Specifications
Specifications
Safety lancet
Lancet for blood testing
Medical sharp
100 ct. per box