Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ergodyne™ ProFlex™ 100HV Economy Spandex Hi-Vis Back Support Brace
$36.75 - $270.50
Specifications
Color | Hi-Vis Orange |
---|---|
Closure Type | Adjustable two stage |
Product Type | Economy Back Support |
For Use With (Application) | Material handling, construction, warehousing/ distribution, maintenance, freight/baggage, assembly/fabrication, landscaping/grounds |
Includes | Non-conductive Polypropylene stays, hi-vis power panels, 1.25 in. adjustable knit suspenders, stretchable bias binding with zig-zag stitching |
Catalog Number | Mfr. No. | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Size | Price | Quantity & Availability | |||||
19-039-199C
|
Ergodyne™
11884 |
Large |
Case of 6 for $270.50
|
|
|||||
19-039-199D
|
Ergodyne™
11885 |
X-Large |
Each for $36.75
|
|
|||||
19-039-199B
|
Ergodyne™
11883 |
Medium |
Each for $36.75
|
|
|||||
Specifications
Hi-Vis Orange | |
Economy Back Support | |
Non-conductive Polypropylene stays, hi-vis power panels, 1.25 in. adjustable knit suspenders, stretchable bias binding with zig-zag stitching | |
100HV | |
Tracked |
Adjustable two stage | |
Material handling, construction, warehousing/ distribution, maintenance, freight/baggage, assembly/fabrication, landscaping/grounds | |
280D Spandex | |
Rubber |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title