Learn More
GSC Go Science Crazy Equilateral Prism Glass
Prisms are a great teaching tool for demonstrations using light. The GSC Equilateral Prism Glass is a great teaching tool for demonstrations using light. This prism is right angle and glass.
Supplier: GSC Go Science Crazy 49097010
Alert:
Not a toy. Product intended for ages 13 and up.
Description
- Optically clear
- Glass
- Equilateral
- Individually boxed
Specifications
Glass | |
Equilateral | |
Prism | |
50 mm |