Learn More
Mti Corporation Quartz Boat 115 mm L x 40 mm W x 20 mm
SDP
Supplier:  Mti Corporation EQQB1042
Encompass
Quartz boat is made of ultra-high purity quartz (4N purity) with 1200oC working temperatureDimension: 115 mm L x 40 mm W x 20 mm HCan fit in MTI 50 - 80 mm O.D tube furnace
Catalog No. NC0782224