Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Eppendorf™ 5810 Series Four- Place Swinging Bucket Tube Adapters
Adapter for the A-4-62 rotor
Supplier: Eppendorf™ 022638203
Description
- Capacity: 1.2 to 5mL tubes
- Maximum tube length with or without cap: 115 to 123mm
- Bore diameter: 11mm
Specifications
A-4-62 swing bucket rotor for 5810/5810R centrifuge | |
1.1 cm |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction