Learn More
Eppendorf™ 18-Place Fixed-Angle Rotor with Cover
For use with discontinued Model 5415C Microcentrifuge
Supplier: Eppendorf™ 022366726
Description
18 × 1.5 to 2.0mL capacity. Also accepts smaller tube sizes when equipped with accessory tube adapters (below). For aerosol containment, this rotor can be used with aerosol containment capsules. (Ask your Fisher Scientific™ Customer Service Representative for details about Aerosol Containment Kit, Cat. No. 05-406-4.) Reaches speeds up to 14,000rpm/16,000×G.
Specifications
Fixed Angle Rotor | |
16,000 x g | |
Eppendorf 5415C Microcentrifuge | |
Lid |
18 x 1.5/2 mL | |
14,000 rpm | |
Fixed |