Learn More
Corning™ Epidermal Growth Factor (EGF), Mouse Natural (Receptor Grade)
Corning™ Epidermal Growth Factor (EGF), mouse natural (receptor grade) is a low-molecular-weight mitogenic protein that stimulates proliferation of a wide variety of cell types in vitro. These growth factors have been used to promote long-term proliferation in cell culture and to induce cell differentiation in a variety of cell types under serum-free or reduced serum conditions.
Supplier: Corning™ 356010
Description
Formulation
Lyophilized from ammonium acetate, carrier-free; reconstitute in dH2O
Preparation and Storage
Stable for at least three months at 2° to 8°C (lyophilized) or =one month solubilized at -20°C . Do not store in frost-free freezer. Avoid multiple freeze/thaws.
Quality
- Purity: >98% by SDS-PAGE and pH 9.9 PAGE
- Tested for ability to stimulate proliferation of human foreskin fibroblasts in serum-containing medium
- Filtered (0.2 μm membrane) and tested and found negative for bacteria, fungi, and mycoplasma
Source
Male mouse submaxillary glands
Molecular Weight
6.1 kD
Purification
EGF, mouse natural (receptor grade) - Purified by BioGel™ P10/DEAE chromatography
Tip
Recommended concentration: Approximately 5 ng/ mL medium depending on cell type
Storage
Shelf life: ≥ 3 months from date of shipment at 2-8°C (lyophilized) or ≥ one month at -20°C (solubilized)
Specifications
Do not store in frost-free freezer. Avoid multiple freeze/thaws. | |
500 μg | |
Mouse |
>98% by SDS-PAGE | |
Male mouse submaxillary glands |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.