Learn More
EPA 8011 EDB/DBCP Mix, Certified Reference Material, 2,000 μg/mL Each Component in Methanol, MilliporeSigma™ Supelco™
This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.
Manufacturer:  MilliporeSigma Supelco 48225U
Catalog No. 11-101-7193
Add to cart
Description
Description
Specifications
Specifications
1,2-Dibromo-3-chloropropane; 1,2-Dibromoethane
TraceCERT™ Analytical Standard
1 mL
Organic Pollutant Standards
2,000 µg/mL each component in methanol
Ampule
TraceCERT
SDS
Documents
Product Certifications
Promotions
Promotions
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC