Learn More
New England Biolabs, Inc. EnGen sgRNA Synthesis Kit, S. pyogenes – 20 reactions
Supplier: New England Biolabs, Inc. E3322S
The EnGen sgRNA Synthesis Kit, S. pyogenes provides a simple and quick method for transcribing high yields of sgRNA in a single 30-minute reaction, using the supplied reagents and target-specific DNA oligos designed by the user.