Learn More
Lonza Walkersville Endotoxin
Supplier: Lonza Walkersville 00193783
Product 00193783 contains high potency endotoxin and is intended for use in endotoxin removal system challenges, i.e. depyrogenation ovens, and other spiking studies. Benefits:Does not contain fillerApplications:For oven validation studiesContent:> 1.25 million EU/mL